Anti-COPS5 Mouse Monoclonal Antibody [clone: OTI2B12]

Proveedor: OriGene
ORIGTA504344EA 608 EUR
ORIGTA504344
Anti-COPS5 Mouse Monoclonal Antibody [clone: OTI2B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR