Anti-MMACHC Mouse Monoclonal Antibody [clone: OTI4E9]

Proveedor: OriGene
ORIGTA504343EA 581 EUR
ORIGTA504343
Anti-MMACHC Mouse Monoclonal Antibody [clone: OTI4E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MMACHC (methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR