Anti-FXN Mouse Monoclonal Antibody [clone: OTI3F3]

Proveedor: OriGene
ORIGTA504341EA 603 EUR
ORIGTA504341
Anti-FXN Mouse Monoclonal Antibody [clone: OTI3F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: FXN (frataxin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR