Anti-NPTN Mouse Monoclonal Antibody [clone: OTI3C4]

Proveedor: OriGene
ORIGTA504319EA 608 EUR
ORIGTA504319
Anti-NPTN Mouse Monoclonal Antibody [clone: OTI3C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NPTN (neuroplastin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR