Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1C5]
Proveedor: OriGene
ORIGTA504300EA 581 EUR
ORIGTA504300
Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: EPN2 (epsin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now