Anti-FMR1 Mouse Monoclonal Antibody [clone: OTI1D10]

Proveedor: OriGene
ORIGTA504291EA 603 EUR
ORIGTA504291
Anti-FMR1 Mouse Monoclonal Antibody [clone: OTI1D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:50

Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR