Anti-RFXANK Mouse Monoclonal Antibody [clone: OTI3F5]
Proveedor: OriGene
ORIGTA504270EA 603 EUR
ORIGTA504270
Anti-RFXANK Mouse Monoclonal Antibody [clone: OTI3F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: RFXANK (regulatory factor X-associated ankyrin-containing protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human