Anti-C1S Mouse Monoclonal Antibody [clone: OTI5F2]

Proveedor: OriGene
ORIGTA504245EA 581 EUR
ORIGTA504245
Anti-C1S Mouse Monoclonal Antibody [clone: OTI5F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR