Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI1G3]

Proveedor: OriGene
ORIGTA504197EA 603 EUR
ORIGTA504197
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI1G3]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KCNJ3 (potassium inwardly-rectifying channel, subfamily J, member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR