Anti-AGPAT5 Mouse Monoclonal Antibody [clone: OTI4H3]

Proveedor: OriGene
ORIGTA504145EA 603 EUR
ORIGTA504145
Anti-AGPAT5 Mouse Monoclonal Antibody [clone: OTI4H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: AGPAT5 (1-acylglycerol-3-phosphate O-acyltransferase 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR