Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI1D3]

Proveedor: OriGene
ORIGTA504139EA 603 EUR
ORIGTA504139
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI1D3]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: KCNJ3 (potassium inwardly-rectifying channel, subfamily J, member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR