Anti-SERPINB6 Mouse Monoclonal Antibody [clone: OTI1C5]

Proveedor: OriGene
ORIGTA504127EA 603 EUR
ORIGTA504127
Anti-SERPINB6 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINB6 (serpin peptidase inhibitor, clade B (ovalbumin), member 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR