Anti-RNF113B Mouse Monoclonal Antibody [clone: OTI2H5]

Proveedor: OriGene
TA504118
ORIGTA504118EA 616 EUR
ORIGTA504118
Anti-RNF113B Mouse Monoclonal Antibody [clone: OTI2H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF113B (ring finger protein 113B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR