Anti-GSTO2 Mouse Monoclonal Antibody [clone: OTI4H1]

Proveedor: OriGene
ORIGTA504085EA 603 EUR
ORIGTA504085
Anti-GSTO2 Mouse Monoclonal Antibody [clone: OTI4H1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GSTO2 (glutathione S-transferase omega 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR