Anti-DGKA Mouse Monoclonal Antibody [clone: OTI6A11]

Proveedor: OriGene
ORIGTA504050EA 603 EUR
ORIGTA504050
Anti-DGKA Mouse Monoclonal Antibody [clone: OTI6A11]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DGKA (diacylglycerol kinase, alpha 80kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR