Anti-SERPINB2 Mouse Monoclonal Antibody [clone: OTI4A3]

Proveedor: OriGene
ORIGTA504003EA 616 EUR
ORIGTA504003
Anti-SERPINB2 Mouse Monoclonal Antibody [clone: OTI4A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINB2 (serpin peptidase inhibitor, clade B (ovalbumin), member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR