Anti-KCNAB1 Mouse Monoclonal Antibody [clone: OTI9F7]

Proveedor: OriGene
ORIGTA503967EA 603 EUR
ORIGTA503967
Anti-KCNAB1 Mouse Monoclonal Antibody [clone: OTI9F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: KCNAB1 (potassium voltage-gated channel, shaker-related subfamily, beta member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR