Anti-TAL1 Mouse Monoclonal Antibody [clone: OTI6A2]

Proveedor: OriGene
ORIGTA503921EA 603 EUR
ORIGTA503921
Anti-TAL1 Mouse Monoclonal Antibody [clone: OTI6A2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TAL1 (T-cell acute lymphocytic leukemia 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR