Anti-NDUFA5 Mouse Monoclonal Antibody [clone: OTI1E8]
Proveedor: OriGene
ORIGTA503881EA 608 EUR
ORIGTA503881
Anti-NDUFA5 Mouse Monoclonal Antibody [clone: OTI1E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: NDUFA5 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human