Anti-CYP2A6 Mouse Monoclonal Antibody [clone: OTI2A8]

Proveedor: OriGene
ORIGTA503827EA 603 EUR
ORIGTA503827
Anti-CYP2A6 Mouse Monoclonal Antibody [clone: OTI2A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP2A6 (cytochrome P450, family 2, subfamily A, polypeptide 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR