Anti-LPL Mouse Monoclonal Antibody [clone: OTI4G2]

Proveedor: OriGene
ORIGTA503810EA 608 EUR
ORIGTA503810
Anti-LPL Mouse Monoclonal Antibody [clone: OTI4G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LPL (lipoprotein lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR