Anti-METTL21A Mouse Monoclonal Antibody [clone: OTI2D3]

Proveedor: OriGene
ORIGTA503803EA 603 EUR
ORIGTA503803
Anti-METTL21A Mouse Monoclonal Antibody [clone: OTI2D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: METTL21A (methyltransferase like 21A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR