Anti-EFEMP1 Mouse Monoclonal Antibody [clone: OTI1D9]

Proveedor: OriGene
ORIGTA503772EA 608 EUR
ORIGTA503772
Anti-EFEMP1 Mouse Monoclonal Antibody [clone: OTI1D9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: EFEMP1 (EGF containing fibulin-like extracellular matrix protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR