Anti-TBC1D21 Mouse Monoclonal Antibody [clone: OTI1A8]

Proveedor: OriGene
ORIGTA503741EA 581 EUR
ORIGTA503741
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: OTI1A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TBC1D21 (TBC1 domain family, member 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR