Anti-ASCC1 Mouse Monoclonal Antibody [clone: OTI2D7]

Proveedor: OriGene
ORIGTA503668EA 603 EUR
ORIGTA503668
Anti-ASCC1 Mouse Monoclonal Antibody [clone: OTI2D7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ASCC1 (activating signal cointegrator 1 complex subunit 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR