Anti-PSMB4 Mouse Monoclonal Antibody [clone: OTI1F8]

Proveedor: OriGene
ORIGTA503637EA 616 EUR
ORIGTA503637
Anti-PSMB4 Mouse Monoclonal Antibody [clone: OTI1F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PSMB4 (proteasome (prosome, macropain) subunit, beta type, 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR