Anti-PRMT2 Mouse Monoclonal Antibody [clone: OTI5F9]

Proveedor: OriGene
ORIGTA503587EA 581 EUR
ORIGTA503587
Anti-PRMT2 Mouse Monoclonal Antibody [clone: OTI5F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRMT2 (protein arginine methyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR