Anti-SEC14L2 Mouse Monoclonal Antibody [clone: OTI2F9]

Proveedor: OriGene
TA503576
ORIGTA503576EA 616 EUR
ORIGTA503576
Anti-SEC14L2 Mouse Monoclonal Antibody [clone: OTI2F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SEC14L2 (SEC14-like 2 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR