Anti-EIF2B3 Mouse Monoclonal Antibody [clone: OTI2D2]

Proveedor: OriGene
TA503568
ORIGTA503568EA 616 EUR
ORIGTA503568
Anti-EIF2B3 Mouse Monoclonal Antibody [clone: OTI2D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: EIF2B3 (eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR