Anti-ELK3 Mouse Monoclonal Antibody [clone: OTI2G5]

Proveedor: OriGene
ORIGTA503560EA 603 EUR
ORIGTA503560
Anti-ELK3 Mouse Monoclonal Antibody [clone: OTI2G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ELK3 (ELK3, ETS-domain protein (SRF accessory protein 2))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR