Anti-WWTR1 Mouse Monoclonal Antibody [clone: OTI1H9]

Proveedor: OriGene
ORIGTA503538EA 581 EUR
ORIGTA503538
Anti-WWTR1 Mouse Monoclonal Antibody [clone: OTI1H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: WWTR1 (WW domain containing transcription regulator 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR