Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI1D12]

Proveedor: OriGene
ORIGTA503471EA 608 EUR
ORIGTA503471
Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI1D12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B (phosphodiesterase 4B, cAMP-specific)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR