Anti-CYP17A1 Mouse Monoclonal Antibody [clone: OTI3D9]

Proveedor: OriGene
ORIGTA503437EA 603 EUR
ORIGTA503437
Anti-CYP17A1 Mouse Monoclonal Antibody [clone: OTI3D9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CYP17A1 (cytochrome P450, family 17, subfamily A, polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR