Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: OTI6G10]

Proveedor: OriGene
ORIGTA503429EA 581 EUR
ORIGTA503429
Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: OTI6G10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMPRSS5 (transmembrane protease, serine 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR