Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI2H10]

Proveedor: OriGene
ORIGTA503426EA 603 EUR
ORIGTA503426
Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI2H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PGAM2 (phosphoglycerate mutase 2 (muscle))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR