Anti-CCM2 Mouse Monoclonal Antibody [clone: OTI3B7]

Proveedor: OriGene
ORIGTA503397EA 603 EUR
ORIGTA503397
Anti-CCM2 Mouse Monoclonal Antibody [clone: OTI3B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CCM2 (cerebral cavernous malformation 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR