Anti-DLD Mouse Monoclonal Antibody [clone: OTI8A10]

Proveedor: OriGene
ORIGTA503388EA 603 EUR
ORIGTA503388
Anti-DLD Mouse Monoclonal Antibody [clone: OTI8A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: DLD (dihydrolipoamide dehydrogenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR