Anti-SETD7 Mouse Monoclonal Antibody [clone: OTI2E3]

Proveedor: OriGene
ORIGTA503377EA 603 EUR
ORIGTA503377
Anti-SETD7 Mouse Monoclonal Antibody [clone: OTI2E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SETD7 (SET domain containing (lysine methyltransferase) 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR