Anti-ATG3 Mouse Monoclonal Antibody [clone: OTI4F6]

Proveedor: OriGene
ORIGTA503370EA 603 EUR
ORIGTA503370
Anti-ATG3 Mouse Monoclonal Antibody [clone: OTI4F6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATG3 (autophagy related 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR