Anti-TMOD1 Mouse Monoclonal Antibody [clone: OTI1H10]

Proveedor: OriGene
ORIGTA503366EA 603 EUR
ORIGTA503366
Anti-TMOD1 Mouse Monoclonal Antibody [clone: OTI1H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TMOD1 (tropomodulin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR