Anti-NDEL1 Mouse Monoclonal Antibody [clone: OTI5E11]

Proveedor: OriGene
ORIGTA503353EA 603 EUR
ORIGTA503353
Anti-NDEL1 Mouse Monoclonal Antibody [clone: OTI5E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NDEL1 (nudE neurodevelopment protein 1-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR