Anti-DOK2 Mouse Monoclonal Antibody [clone: OTI3D6]

Proveedor: OriGene
ORIGTA503337EA 603 EUR
ORIGTA503337
Anti-DOK2 Mouse Monoclonal Antibody [clone: OTI3D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DOK2 (docking protein 2, 56kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR