Anti-PDE1B Mouse Monoclonal Antibody [clone: OTI1D6]

Proveedor: OriGene
ORIGTA503336EA 603 EUR
ORIGTA503336
Anti-PDE1B Mouse Monoclonal Antibody [clone: OTI1D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PDE1B (phosphodiesterase 1B, calmodulin-dependent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR