Anti-SSR1 Mouse Monoclonal Antibody [clone: OTI3E5]

Proveedor: OriGene
ORIGTA503263EA 581 EUR
ORIGTA503263
Anti-SSR1 Mouse Monoclonal Antibody [clone: OTI3E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SSR1 (signal sequence receptor, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR