Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI1H9]

Proveedor: OriGene
ORIGTA503250EA 603 EUR
ORIGTA503250
Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI1H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY1 (aminoacylase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR