Anti-RLBP1 Mouse Monoclonal Antibody [clone: OTI1B4]

Proveedor: OriGene
ORIGTA503148EA 581 EUR
ORIGTA503148
Anti-RLBP1 Mouse Monoclonal Antibody [clone: OTI1B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RLBP1 (retinaldehyde binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR