Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI1B6]

Proveedor: OriGene
ORIGTA503119EA 603 EUR
ORIGTA503119
Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI1B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TOMM34 (translocase of outer mitochondrial membrane 34)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR