Anti-TUBB4A Mouse Monoclonal Antibody [clone: OTI8E3]

Proveedor: OriGene
ORIGTA503110EA 603 EUR
ORIGTA503110
Anti-TUBB4A Mouse Monoclonal Antibody [clone: OTI8E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TUBB4A (tubulin, beta 4A class IVa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR