Anti-SMS Mouse Monoclonal Antibody [clone: OTI5F2]

Proveedor: OriGene
ORIGTA503098EA 603 EUR
ORIGTA503098
Anti-SMS Mouse Monoclonal Antibody [clone: OTI5F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: SMS (spermine synthase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR