Anti-ITFG2 Mouse Monoclonal Antibody [clone: OTI3A3]

Proveedor: OriGene
ORIGTA503047EA 586 EUR
ORIGTA503047
Anti-ITFG2 Mouse Monoclonal Antibody [clone: OTI3A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: ITFG2 (integrin alpha FG-GAP repeat containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR